Rasfonin ≥95% (by HPLC)

Danger

Synonyms: [(2R,3R)-2-[(E,2S,4R)-4,6-dimethyloct-6-en-2-yl]-6-oxo-2,3-dihydropyran-3-yl] (2E,4E,6S)-8-hydroxy-6-(hydroxymethyl)-4-methylocta-2,4-dienoate

B1977-500
76268-852EA 262.69 USD
76268-852
Rasfonin ≥95% (by HPLC)
Rasfonin

Rasfonin, isolated from Talaromyces sp., is a selective apoptosis inducer in ras-dependent cells. Rasfonin induces autophagy concomitant with upregulation of Akt phosphorylation.

Formula: C₂₅H₃₈O₆
MW: 434,57 g/mol
CAS Number: 303156-68-5
https://us.vwr.com/stibo/web/std.lang.all/02/54/24140254.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR