BIOVB1973
B1973-25 B1973-5
76268-762EA 180.25 USD
76268-762 76268-764
BIOVB1973
AKP-11 is a novel S1P1 (Sphingosine-1-phosphate receptor 1) agonist with potential therapeutic potential for treatment of MS and related autoimmune disorders.