IPI-549 ≥98% (by HPLC)

Synonyms: 2-amino-N-[1-[8-[2-(1-methylpyrazol-4-yl)ethynyl]-1-oxo-2-phenylisoquinolin-3-yl]ethyl]pyrazolo[1,5-a]pyrimidine-3-carboxamide(S)-2-Amino-N-(1-(8-((1-methyl-1H-pyrazol-4-yl)ethynyl)-1-oxo-2-phenyl-1,2-dihydroisoquinolin-3-yl)ethyl)pyrazolo[1,5-a]pyrimidine-3-carboxamide

B1971-25 B1971-5
76268-754EA 134.25 USD
76268-754 76268-756
IPI-549 ≥98% (by HPLC)
IPI-549

IPI-549 is a potent and selective PI3Kγ Inhibitor (IC₅₀ of 16 nM). It displays no significant inhibition for a panel of 468 mutant and non-mutant protein and lipid kinases (including Class II PI3K isoforms) at 1 µM.

Formula: C₃₀H₂₄N₈O₂
MW: 528,56 g/mol
MDL Number: MFCD30533720
CAS Number: 1693758-51-8
https://us.vwr.com/stibo/web/std.lang.all/02/48/24140248.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR