MTX-211 ≥98% (by HPLC)

Synonyms: N-[2-chloro-5-[4-(3-chloro-4-fluoroanilino)quinazolin-6-yl]pyridin-3-yl]methanesulfonamideN-[2-chloro-5-[4-(3-chloro-4-fluoroanilino)quinazolin-6-yl]pyridin-3-yl]methanesulphonamide

B1968-5 B1968-25
76268-744EA 577.91 USD
76268-744 76268-742
MTX-211 ≥98% (by HPLC)
MTX-211

MTX-211 is a dual inhibitor of EGFR and PI3K. Potentially useful for treatment of KRAS mutant colorectal cancer.

Formula: C₂₀H₁₄Cl₂FN₅O₂S
MW: 478,33 g/mol
CAS Number: 1952236-05-3
https://us.vwr.com/stibo/web/std.lang.all/02/46/24140246.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR