DDP-38003 dihydrochloride ≥98% (by HPLC)

Warning

Synonyms: dihydrochlorideN-[4-[(1R,2S)-2-aminocyclopropyl]cyclohexyl]-4-(4-methylpiperazin-1-yl)benzamide

B1958-5 B1958-25
76268-722EA 204.31 USD
76268-722 76268-724
DDP-38003 dihydrochloride ≥98% (by HPLC)
DDP-38003 dihydrochloride

DDP-38003 dihydrochloride is a potent, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with an IC₅₀ of 84 nM.

Formula: C₂₁H₂₈Cl₂N₄O
MW: 423,38 g/mol
CAS Number: 1831167-98-6
https://us.vwr.com/stibo/web/std.lang.all/02/22/24140222.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR