ORY-1001 ≥98% (by HPLC)

Warning

Synonyms: N1-((1R,2S)-2-phenylcyclopropyl)cyclohexane-1,4-diaminedihydrochloridetrans-N1-(2-Phenylcyclopropyl)cyclohexane-1,4-diamine dihydrochlorideRG-6016

B1957-5 B1957-1
76268-720EA 437.81 USD
76268-720 76268-718
ORY-1001 ≥98% (by HPLC)
ORY-1001

ORY-1001 is a potent and selective inhibitor of LSD1 (IC₅₀ < 20 nM). It targets acute myeloid leukemia (AML) stem cells and significantly reduces tumor cell load while increasing survival time in mouse models of AML. ORY-1001 is in clinical trials for cancer treatment.

Formula: C₁₅H₂₄Cl₂N₂
MW: 303,27 g/mol
MDL Number: MFCD28900684
CAS Number: 1431326-61-2
https://us.vwr.com/stibo/web/std.lang.all/02/21/24140221.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR