NCGC00244536 ≥98% (by HPLC)

Synonyms: 3-(8-hydroxyquinolin-6-yl)-N-(3-phenylpropyl)benzamide

B1956-5 B1956-25
76268-714EA 145.93 USD
76268-714 76268-716
NCGC00244536 ≥98% (by HPLC)
NCGC00244536

NCGC00244536 is a potent KDM4A inhibitor (IC₅₀ = 10 nM). NCGC00244536 displays high selectivity for the fast-growing AR-negative PC3 cells (IC₅₀ = 40 nM) and over 100-fold selectivity against the immortalized prostate epithelial cell lines PrEC1 and PrEC4. Displays anticancer activity.

Formula: C₂₅H₂₂N₂O₂
MW: 382,46 g/mol
MDL Number: MFCD31382180
CAS Number: 2003260-55-5
https://us.vwr.com/stibo/web/std.lang.all/02/20/24140220.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR