Ac-DEVD-CHO
Supplier: BioVision
B1951-5 B1951-1
76268-696EA 391.11 USD
76268-696 76268-694
Ac-DEVD-CHO
Proteins and Peptides
Ac-DEVD-CHO is a potent and reversible inhibitor of the Group II caspases, caspase-3 and caspase-7 (Ki's = 0.23 and 1.6 nM, respectively). The DEVD sequence is based on PARP cleavage site Asp-216 for caspase-3.
  • A reversible caspase-3 inhibitor