A-1331852 ≥98% (by HPLC)

Synonyms: 3-(1-(adamantan-1-ylmethyl)-5-methyl-1H-pyrazol-4-yl)-6-(8-(benzo[d]thiazol-2-ylcarbamoyl)-3,4-dihydroisoquinolin-2(1H)-yl) picolinic acid

B1948-5 B1948-25
76268-684EA 758.88 USD
76268-684 76268-682
A-1331852 ≥98% (by HPLC)
A-1331852

A-1331852 is a potent anti-apoptotic Bcl-xL protein inhibitor and apoptosis-inducing agent useful in the treatment of cancer, immune and autoimmune diseases.

Formula: C₃₈H₃₈N₆O₃S
MW: 658,81 g/mol
CAS Number: 1430844-80-6
https://us.vwr.com/stibo/web/std.lang.all/02/11/24140211.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR