Ensartinib ≥98% (by HPLC)

Synonyms: (R)-6-Amino-5-(1-(2,6-dichloro-3-fluorophenyl)ethoxy)-N-(4-(4-methylpiperazine-1-carbonyl)phenyl)pyridazine-3-carboxamide

B1946-25 B1946-5
76268-674EA 192.64 USD
76268-674 76268-676
Ensartinib ≥98% (by HPLC)
Ensartinib

Ensartinib (X-396), is a potent, small molecule inhibitor of the receptor tyrosine kinase anaplastic lymphoma kinase (ALK) with potential antineoplastic activity. Ensartinib has been active in animal models of non-small cell lung cancer (NSCLC) and neuroblastoma. Also, ensartinib has shown activity in models with ALK mutations that confer resistance to other small molecule ALK inhibitors.

Formula: C₂₅H₂₅Cl₂FN₆O₃
MW: 547,41 g/mol
MDL Number: MFCD25976868
CAS Number: 1365267-27-1 
https://us.vwr.com/stibo/web/std.lang.all/02/08/24140208.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR