TAK-901 ≥98% (by HPLC)

Synonyms: 5-(3-ethylsulphonylphenyl)-3,8-dimethyl-N-(1-methylpiperidin-4-yl)-9H-pyrido[2,3-b]indole-7-carboxamide5-(3-ethylsulfonylphenyl)-3,8-dimethyl-N-(1-methylpiperidin-4-yl)-9H-pyrido[2,3-b]indole-7-carboxamide

B1945-5 B1945-25
76268-850EA 846.44 USD
76268-850 76268-848
TAK-901 ≥98% (by HPLC)
TAK-901

TAK-901 is a potent inhibitor of Aurora kinases A and B with IC₅₀ value of 21 nM and 15 nM, respectively. Displays antineoplastic activity.

Formula: C₂₈H₃₂N₄O₃S
MW: 504,64 g/mol
CAS Number: 934541-31-8
https://us.vwr.com/stibo/web/std.lang.all/02/07/24140207.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR