Hesperadin ≥98% (by HPLC)

Synonyms: (Z)-N-(2-Oxo-3-(phenyl((4-(piperidin-1-ylmethyl)phenyl)amino)methylene)indolin-5-yl)ethanesulphonamide(Z)-N-[2-Oxo-3-[phenyl[[4-(piperidin-1-ylmethyl)phenyl]amino]methylene]indolin-5-yl]ethanesulphonamide(Z)-N-[2-Oxo-3-[phenyl[[4-(piperidin-1-ylmethyl)phenyl]amino]methylene]indolin-5-yl]ethanesulfonamide(Z)-N-(2-Oxo-3-(phenyl((4-(piperidin-1-ylmethyl)phenyl)amino)methylene)indolin-5-yl)ethanesulfonamide

B1940-25 B1940-5
76268-656EA 215.99 USD
76268-656 76268-658
Hesperadin ≥98% (by HPLC)
Hesperadin

Hesperadin is a potent, ATP-competitive inhibitor of Aurora B kinase (IC₅₀ = 250 nM). Hesperadin inhibits Aurora B kinase by phosphorylation of histone H3. Also acts as a broad-spectrum influenza antiviral agent.

Formula: C₂₉H₃₂N₄O₃S
MW: 516,65 g/mol
MDL Number: MFCD18074526
CAS Number: 422513-13-1
https://us.vwr.com/stibo/web/std.lang.all/02/01/24140201.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR