GSK1070916
B1939-5 B1939-25
76268-652EA 192.64 USD
76268-652 76268-654
GSK1070916
GSK1070916
GSK-1070916 is a potent, reversible and ATP-competitive inhibitor of Aurora kinases B and C with IC₅₀ value of 3.5 nM and 6.5 nM, respectively. Treatment of GSK-1070916 suppresses cell proliferation in A549 human lung cancer cells with EC₅₀ value of 7 nM.
CAS Number: 942918-07-2
https://us.vwr.com/stibo/web/std.lang.all/01/99/24140199.jpg