CCT-129202 ≥98% (by HPLC)

Synonyms: 2-[4-[6-chloro-2-[4-(dimethylamino)phenyl]-1H-imidazo[4,5-b]pyridin-7-yl]piperazin-1-yl]-N-(1,3-thiazol-2-yl)acetamide

B1934-1 B1934-5
76268-814EA 99.24 USD
76268-814 76268-836
CCT-129202 ≥98% (by HPLC)
CCT-129202

CCT-129202 is a potent, ATP-competitive inhibitor of Aurora kinases with IC₅₀ values of 42 nM, 198 nM and 227 nM for Aurora A, B and C, respectively. CCT-129202 displays high selectivity for the Aurora kinases over a panel of other kinases tested and inhibits proliferation in multiple cultured human tumor cell lines.

Formula: C₂₃H₂₅ClN₈OS
MW: 497,02 g/mol
CAS Number: 942947-93-5
https://us.vwr.com/stibo/web/std.lang.all/01/92/24140192.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR