DREADD Agonist 21 ≥98% (by HPLC)

Supplier: BioVision

Synonyms: 11-(1-Piperazinyl)-5H-dibenzo[b,e][1,4]diazepine

AB235545-5MG AB235545-1MG
76268-574EA 574.58 USD
76268-574 76268-572
DREADD Agonist 21 ≥98% (by HPLC)
DREADD Agonist 21

DREADD agonist 21 is a potent muscarinic hM3Dq DREADD agonist (EC₅₀ = 1.7 nM).

Formula: C₁₇H₁₈N₄
MW: 278,35 g/mol
CAS Number: 56296-18-5
https://us.vwr.com/stibo/web/std.lang.all/01/77/24140177.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR