OSMI-1 ≥98% (by HPLC)

Supplier: BioVision

Synonyms: OSMI 1αR-α-[[(1,2-Dihydro-2-oxo-6-quinolinyl)sulphonyl]amino]-N-(2-furanylmethyl)-2-methoxy-N-(2-thienylmethyl)-benzeneacetamideαR-α-[[(1,2-Dihydro-2-oxo-6-quinolinyl)sulfonyl]amino]-N-(2-furanylmethyl)-2-methoxy-N-(2-thienylmethyl)-benzeneacetamide

AB235455-25MG
76268-864EA 283.46 USD
76268-864 76268-866
OSMI-1 ≥98% (by HPLC)
OSMI-1

OSMI-1 is a potent, cell-permeable, O-GlcNAc transferase (OGT) inhibitor.  inhibits protein O-GlcNAcylation in several mammalian cell lines without qualitatively altering cell surface N- or O-linked glycans.

Formula: C₂₈H₂₅N₃O₆S₂
MW: 563,64 g/mol
MDL Number: MFCD30187585
CAS Number: 1681056-61-0
https://us.vwr.com/stibo/web/std.lang.all/01/65/24140165.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR