Pimavanserin Tartrate ≥97%

Danger

Synonyms: 1-(4-fluorobenzyl)-3-(4-isobutoxybenzyl)-1-(1-methylpiperidin-4-yl)urea hemi((2R,3R)-2,3-dihydroxysuccinate)ACP-103

B1915-50 B1915-10
76270-338EA 391.11 USD
76270-338 76270-336
Pimavanserin Tartrate ≥97%
Pimavanserin Tartrate

Pimavanserin tartrate is an inverse agonist on the serotonin receptor subtype 5-HT2A, that reverses psychosis-like behavior and has the potential to treat various other neuropsychiatric disorders such as schizophrenia and Parkinson’s disease.

Formula: C₅₄H₇₄F₂N₆O₁₀
MW: 1005,21 g/mol
CAS Number: 706782-28-7
https://us.vwr.com/stibo/web/std.lang.all/01/64/24140164.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR