Tanshinone IIA-sulfonic sodium ≥98% (by HPLC)

Synonyms: Sodium 1,6,6-trimethyl-10,11-dioxo-6,7,8,9,10,11-hexahydrophenanthro[1,2-b]furan-2-sulphonatesulfotanshinone sodium II-Asulphotanshinone sodium II-ASodium tanshinone IIA sulfonateSodium 1,6,6-trimethyl-10,11-dioxo-6,7,8,9,10,11-hexahydrophenanthro[1,2-b]furan-2-sulfonateT06Sodium tanshinone IIA sulphonate

B1902-5 B1902-25
76270-238EA 332.74 USD
76270-238 76270-236
Tanshinone IIA-sulfonic sodium ≥98% (by HPLC)
Tanshinone IIA-sulfonic sodium

Sodium tanshinone IIA sulphonate (T06) is a water-soluble derivative of tanshinone IIA, that displays versatile biological properties. Recently, it has been shown to act as an ectosteric inhibitor of cathepsin K (CatK). T06 binds in an ectosteric side of Cathepsin K remote from its active site and selectively inhibits collagen degradation. It also inhibits bone resorption in ovariectomized mice.

Formula: C₁₉H₁₇NaO₆S
MW: 396,39 g/mol
MDL Number: MFCD09028094
CAS Number: 69659-80-9
https://us.vwr.com/stibo/web/std.lang.all/01/44/24140144.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR