Atazanavir

Danger

Synonyms: 1,14-Dimethyl (3S,8S,9S,12S)-3,12-bis(1,1-dimethylethyl)-8-hydroxy-4,11-dioxo-9-(phenylmethyl)-6-[[4-(2-pyridinyl)phenyl]methyl]-2,5,6,10,13-pentaazatetradecanedioate

B1901-5 B1901-25
76270-234EA 531.21 USD
76270-234 76270-232
Atazanavir
Atazanavir

Atazanavir is a potent inhibitor of HIV protease (Ki of 2.66 nM). Atazanavir binds to HIV-1 protease preventing the cleavage of gag and gag-pol polyproteins, which results in the formation of immature virions in HIV-1-infected cells.

Formula: C₃₈H₅₂N₆O₇
MW: 704.87 g/mol
MDL Number: MFCD08435966
CAS Number: 198904-31-3
https://us.vwr.com/stibo/web/std.lang.all/01/43/24140143.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR