Zanamivir ≥98% (by HPLC)

Warning

Synonyms: (2R,3R,4S)-3-Acetamido-4-guanidino-2-((1R,2R)-1,2,3-trihydroxypropyl)-3,4-dihydro-2H-pyran-6-carboxylic acid4-Guanidino-2,4-dideoxy-2,3-dehydro-N-acetylneuraminic acid(2R,3R,4S)-3-acetamido-4-(diaminomethylideneamino)-2-[(1R,2R)-1,2,3-trihydroxypropyl]-3,4-dihydro-2H-pyran-6-carboxylic acid4-Guanidino-Neu5Ac2en

B1900-10 B1900-50
76270-230EA 391.11 USD
76270-230 76270-228
Zanamivir ≥98% (by HPLC)
Zanamivir

Zanamivir is a potent and selective inhibitor of influenza A and B virus neuraminidases with IC₅₀ values of 0.95 nM and 2.7 nM, respectively. It interacts with a group of amino acids in the active site of neuraminidase, that are conserved in all influenza A and B strains. Zanamivir blocks the action of neuraminidase, which blocks the cleavage of sialic acid on the cell receptors, thus preventing release and spread of the newly formed virions.

Formula: C₁₂H₂₀N₄O₇
MW: 332,31 g/mol
MDL Number: MFCD00866966
CAS Number: 139110-80-8
https://us.vwr.com/stibo/web/std.lang.all/01/42/24140142.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR