Oseltamivir acid ≥95%

Synonyms: Oseltamivir carboxylate4R-(acetylamino)-5S-amino-3R-(1-ethylpropoxy)-1-cyclohexene-1-carboxylic acidGS-4071

B1886-1 B1886-5
76270-114EA 321.87 USD
76270-114 76270-112
Oseltamivir acid ≥95%
Oseltamivir acid

Oseltamivir acid (GS-4071) is the active metabolite of oseltamivir (Cat. No. B1885). It acts as an inhibitor of influenza neuraminidases A and B (IC₅₀ = 0.1 to 4.9 nM for both enzymes), in this way preventing virus budding and release.

Formula: C₁₄H₂₄N₂O₄
MW: 284,35 g/mol
MDL Number: MFCD00953940
CAS Number: 187227-45-8
https://us.vwr.com/stibo/web/std.lang.all/01/23/24140123.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR