BI-9564 ≥98% (by HPLC)

Supplier: BioVision

Synonyms: 4-(4-((dimethylamino)methyl)-2,5-dimethoxyphenyl)-2-methyl-2,7-naphthyridin-1(2H)-one

AB254238-5MG AB254238-25MG
76270-088EA 337.09 USD
76270-088 76270-090
BI-9564 ≥98% (by HPLC)
BI-9564

BI-9564 is a potent, selective and cell-permeable BRD9 and BRD7 inhibitor (Kd values are 14 and 239 nM, respectively). Displays selectivity for BRD9/7 over 48 other bromodomains, 324 kinases and 55 GPCRs. Inhibits proliferation of AML cell lines in vitro and reduces AML tumor xenograft growth in vivo.

Formula: C₂₀H₂₃N₃O₃
MW: 353,41 g/mol
MDL Number: MFCD30182324
CAS Number: 1883429-22-8
https://us.vwr.com/stibo/web/std.lang.all/01/14/24140114.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR