L-755,507 ≥98% (by HPLC), EZSolution™

Warning

Synonyms: 4-[[(Hexylamino)carbonyl]amino]-N-[4-[2-[[(2S)-2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino]ethyl]phenyl]-benzenesulfonamide4-[[(Hexylamino)carbonyl]amino]-N-[4-[2-[[(2S)-2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino]ethyl]phenyl]-benzenesulphonamide

B1873-5
76270-040EA 180.96 USD
76270-040
L-755,507 ≥98% (by HPLC), EZSolution™
L-755,507

A 10 mM (5.0 mg in 855 μl), sterile-filtered solution of the HDR (homology-directed repair) enhancer L-755,507 (Cat. No. B1116) in anhydrous DMSO.

Formula: C₃₀H₄₀N₄O₆S
MW: 584,73 g/mol
CAS Number: 159182-43-1
https://us.vwr.com/stibo/web/std.lang.all/01/04/24140104.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR