Brefeldin A 10 mM in DMSO, EZSolution™

Warning

Synonyms: Brefeldin A, natural

B1872-5
76270-038EA 125 USD
76270-038
Brefeldin A 10 mM in DMSO, EZSolution™
Brefeldin A

A 10 mM (5.0 mg in 1.78 ml), sterile-filtered solution of the HDR (homology-directed repair) enhancer Brefeldin A (Cat. No. 1560) in anhydrous DMSO.

Formula: C₁₆H₂₄O₄
MW: 280.4 g/mol
MDL Number: MFCD12913297
CAS Number: 20350-15-6
https://us.vwr.com/stibo/web/std.lang.all/01/03/24140103.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR