BG-45 ≥98% (by HPLC)

Warning

Synonyms: N-(2-Aminophenyl)pyrazine-2-carboxamide

B1866-5 B1866-25
76269-960EA 110.91 USD
76269-960 76269-958
BG-45 ≥98% (by HPLC)
BG-45

BG-45 is a potent HDAC class I inhibitor with selectivity for HDAC3 (IC₅₀ = 289 nM) over HDAC1, 2. BG45 does not inhibit HDAC6. BG-45 alone or in combination with bortezomib has been shown to dose-dependently inhibit tumor growth in a mouse model of multiple myeloma.

Formula: C₁₁H₁₀N₄O
MW: 214,23 g/mol
MDL Number: MFCD09046162
CAS Number: 926259-99-6
https://us.vwr.com/stibo/web/std.lang.all/00/97/24140097.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR