Fulvestrant ≥98% (by HPLC)

Danger

Synonyms: ICI182,780(7R,8R,9S,13S,14S,17S)-13-methyl-7-(9-((4,4,5,5,5-pentafluoropentyl)sulfinyl)nonyl)-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol(7R,8R,9S,13S,14S,17S)-13-methyl-7-(9-((4,4,5,5,5-pentafluoropentyl)sulphinyl)nonyl)-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol

B1863-25 B1863-5
76269-946EA 577.91 USD
76269-946 76269-948
Fulvestrant ≥98% (by HPLC)
Fulvestrant

Fulvestrant is a high affinity estrogen receptor antagonist (IC₅₀ = 0.29 nM), that works by down-regulating and degrading the estrogen receptor, ERα. Reported to lack any partial agonist activity.

Formula: C₂₃H₄₇F₅O₃S
MW: 606,77 g/mol
MDL Number: MFCD00903953
CAS Number: 129453-61-8
https://us.vwr.com/stibo/web/std.lang.all/00/94/24140094.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR