FITM ≥98% (by HPLC)

Synonyms: 4-Fluoro-N-[4-[6-(isopropylamino)pyrimidin-4-yl]-1,3-thiazol-2-yl]-N-methylbenzamide

B1859-5 B1859-1
76269-930EA 110.91 USD
76269-930 76269-932
FITM ≥98% (by HPLC)
FITM

FITM is a potent mGlu1 inhibitor.  It displays high affinity and selectivity for mGlu1 over mGlu5.

Formula: C₁₈H₁₈FN₅OS
MW: 371,43 g/mol
CAS Number: 932737-65-0
https://us.vwr.com/stibo/web/std.lang.all/00/90/24140090.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR