Bosentan ≥98% (by HPLC)
|
Danger
|
Synonyms:
4-(1,1-dimethylethyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]-benzenesulphonamide
, Ro 47-0203
, 4-(1,1-dimethylethyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]-benzenesulfonamide
B1857-1
B1857-5
76269-834EA
87.57
USD
76269-834
76269-836
Bosentan ≥98% (by HPLC)
Bosentan
Bosentan is an antagonist of the endothelin receptors type A (Ki = 6.5 nM) and type B (Ki = 340 nM). Inhibits ET-1 induced contraction of isolated rat aorta and sarafotoxin S6C-induced contraction of rat trachea in vitro.
Formula:
C₂₇H₂₉N₅O₆S MW: 551,61 g/mol |
CAS Number:
147536-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...