U-93631 ≥98% (by HPLC)

Synonyms: 4,5-Dihydro-4,4-dimethyl-imidazo[1,5-a]quinoxaline-3-carboxylic acid 1,1-dimethylethyl estertert-Butyl 4,4-dimethyl-4,5-dihydroimidazo[1,5-a]quinoxaline-3-carboxylate

B1851-25 B1851-5
76269-780EA 531.21 USD
76269-780 76269-778
U-93631 ≥98% (by HPLC)
U-93631

U-93631 is a GABAA receptor antagonist. U-93631 causes rapid decay of gamma-aminobutyric acid-induced chloride currents in recombinant rat gamma-aminobutyric acid type A receptors.

Formula: C₁₇H₂₁N₃O₂
MW: 299,37 g/mol
MDL Number: MFCD10566712
CAS Number: 152273-12-6
https://us.vwr.com/stibo/web/std.lang.all/00/82/24140082.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR