GLPG-1690 ≥98% (by HPLC)

Supplier: BioVision
Danger

Synonyms: 2-((2-ethyl-6-(4-(2-(3-hydroxyazetidin-1-yl)-2-oxoethyl)piperazin-1-yl)-8-methylimidazo[1,2-a]pyridin-3-yl)(methyl)amino)-4-(4-fluorophenyl)thiazole-5-carbonitrile

AB285398-25MG AB285398-5MG
76269-774EA 245.13 USD
76269-774 76269-776
GLPG-1690 ≥98% (by HPLC)
GLPG-1690

GLPG-1690 is a selective autotaxin inhibitor with potential application in idiopathic pulmonary disease (IPF).

Formula: C₃₀H₃₃FN₈O₂S
MW: 588,7 g/mol
MDL Number: MFCD31544330
CAS Number: 1628260-79-6
https://us.vwr.com/stibo/web/std.lang.all/00/81/24140081.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR