RSV-604 ≥98% (by HPLC)

Synonyms: (S)-1-(2-fluorophenyl)-3-(2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)urea

B1846-25 B1846-5
76269-760EA 577.91 USD
76269-760 76269-758
RSV-604 ≥98% (by HPLC)
RSV-604

RSV-604 is a novel inhibitor of respiratory syncytial virus (RSV) replication (EC₅₀ = 0.86 uM). Potentially useful for the treatment of respiratory syncytial virus infections.

Formula: C₂₂H₁₇FN₄O₂
MW: 388,4 g/mol
CAS Number: 676128-63-5
https://us.vwr.com/stibo/web/std.lang.all/00/77/24140077.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR