TAS-103 ≥98% (by HPLC)

Danger

Synonyms: 6-[[2-(Dimethylamino)ethyl]amino]-3-hydroxy-7H-indeno[2,1-c] quinolin-7-one

B1841-25 B1841-5
76269-700EA 145.93 USD
76269-700 76269-702
TAS-103 ≥98% (by HPLC)
TAS-103

TAS-103 (BMS-247615) is a dual inhibitor of topoisomerase-I (topo-I) and topoisomerase-II (topo II).

Formula: C₂₀H₁₉N₃O₂
MW: 333,38 g/mol
CAS Number: 174634-08-3
https://us.vwr.com/stibo/web/std.lang.all/00/72/24140072.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR