ZINC00881524 ≥98% (by HPLC)

Warning

Synonyms: N-(4,5-dihydronaphtho[1,2-d]thiazol-2-yl)-2-(3,4-dimethoxyphenyl)acetamide

B1840-5 B1840-25
76269-696EA 145.89 USD
76269-696 76269-698
ZINC00881524 ≥98% (by HPLC)
ZINC00881524

ZINC00881524 is a potent ROCK (Rho Kinase) inhibitor.

Formula: C₂₁H₂₀N₂O₃S
MW: 380,46 g/mol
MDL Number: MFCD07821731
CAS Number: 557782-81-7
https://us.vwr.com/stibo/web/std.lang.all/00/71/24140071.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR