BMS-986205 ≥98% (by HPLC)

Synonyms: (R)-N-(4-chlorophenyl)-2-((1s,4S)-4-(6-fluoroquinolin-4-yl)cyclohexyl)propenamide(R)-N-(4-Chlorophenyl)-2-(cis-4-(6-fluoroquinolin-4-yl)cyclohexyl)propanamide

B1839-25 B1839-5
76269-692EA 206.79 USD
76269-692 76269-694
BMS-986205 ≥98% (by HPLC)
BMS-986205

BMS-986205 is a potent and selective IDO1 (indoleamine 2,3-dioxygenase 1) inhibitor. Does not have much effect on IDO2 (indoleamine 2,3-dioxygenase 2) or TDO (Tryptophan 2,3-dioxygenase).

Formula: C₂₄H₂₄ClFN₂O
MW: 410,92 g/mol
MDL Number: MFCD31729367
CAS Number: 1923833-60-6
https://us.vwr.com/stibo/web/std.lang.all/00/70/24140070.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR