TCS OX2 29 ≥98% (by HPLC)

Synonyms: (2S)-1-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)-3,3-dimethyl-2-[(4-pyridinylmethyl)amino]-1-butanone hydrochloride

B1838-5 B1838-25
76269-688EA 147.71 USD
76269-688 76269-690
TCS OX2 29 ≥98% (by HPLC)
TCS OX2 29

TCS OX2 29 is a potent and selective OX2 (Orexin-2) receptor antagonist (IC₅₀ = 40 nM). Displays >250-fold selectivity for OX2 over OX1 (Orexin-1) and >50 receptors, ion channels and transporters.

Formula: C₂₃H₃₁N₃O₃.HCl
MW: 433.97 g/mol
CAS Number: 372523-75-6
https://us.vwr.com/stibo/web/std.lang.all/00/69/24140069.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR