PNU-74654

Synonyms: N'-((5-Methylfuran-2-yl)methylene)-2-phenoxybenzohydrazide

B1837-25 B1837-5
76269-678EA 145.93 USD
76269-678 76269-680
PNU-74654
PNU-74654

PNU-74654 is a β-catenin inhibitor (Kd = 450 nM), inhibiting its interaction with the transcription factor T cell factor 4 (Tcf4) resulting in disruption of the Wnt signaling pathway. Treatment of human adrenocortical cell lines with PNU-74654 significantly reduces expression of several Tcf/β-catenin target genes.

Formula: C₁₉H₁₆N₂O₃
MW: 320.3419 g/mol
MDL Number: MFCD12912438
CAS Number: 113906-27-7
https://us.vwr.com/stibo/web/std.lang.all/00/68/24140068.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR