AT-406 ≥98% (by HPLC)

Warning

Synonyms: SM-406(5S,8S,10aR)-N-benzhydryl-5-((S)-2-(methylamino)propanamido)-3-(3-methylbutanoyl)-6-oxodecahydropyrrolo[1,2-a][1,5]diazocine-8-carboxamide

B1829-5 B1829-25
76270-878EA 904.81 USD
76270-878 76270-876
AT-406 ≥98% (by HPLC)
AT-406

AT-406 is a potent inhibitor of IAP (inhibitor of apoptosis protein) family of proteins with potential apoptotic inducing and antineoplastic activity. AT-406 selectively inhibits the biological activity of IAP proteins, including X chromosome-linked IAP (XIAP), the cellular IAPs 1 (c-IAP1) and 2 (c-IAP2) and melanoma inhibitor of apoptosis protein (ML-IAP).

Formula: C₃₂H₄₃N₅O₄
MW: 561,73 g/mol
MDL Number: MFCD22124467
CAS Number: 1071992-99-8
https://us.vwr.com/stibo/web/std.lang.all/00/61/24140061.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR