BDA366 ≥98% (by HPLC)

Warning

Synonyms: 1-[[3-(Diethylamino)-2(S)-hydroxypropyl]amino]-4-[(2(S)-oxiranylmethyl)amino]-9,10-anthracenedione

B1827-25 B1827-5
76270-868EA 157.56 USD
76270-868 76270-870
BDA366 ≥98% (by HPLC)
BDA366

BDA-366 is a small-molecule Bcl2-BH4 domain antagonist, that binds BH4 with high affinity and selectivity. BDA-366-Bcl2 binding induces conformational change in Bcl2 that abrogates its antiapoptotic function, converting it from a survival molecule to a cell death inducer.

Formula: C₂₄H₂₉N₃O₄
MW: 423,5 g/mol
CAS Number: 1909226-00-1
https://us.vwr.com/stibo/web/std.lang.all/00/59/24140059.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR