Meisoindigo ≥98% (by HPLC)

Synonyms: 3-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)-1,3-dihydro-1-methyl-2H-indol-2-one1-Methyl-[3,3'-biindolinylidene]-2,2'-dioneN-Methylisoindigotin

B1822-25 B1822-5
76270-786EA 688.82 USD
76270-786 76270-784
Meisoindigo ≥98% (by HPLC)
Meisoindigo

Meisoindigo is a synthetic derivative of indirubin that acts as a potential agent for acute myeloid leukemia. In the in vitro assay, it can inhibit synthesis of DNA and RNA and the assembly of microtubules. It is also reported to have efficacy in acute myeloid leukemia. In the AML cell lines, HL-60, NB4 and U937, meisoindigo induces apoptosis in both caspase-dependent and -independent pathways.

Formula: C₁₇H₁₂N₂O₂
MW: 276,29 g/mol
MDL Number: MFCD05988503
CAS Number: 97207-47-1
https://us.vwr.com/stibo/web/std.lang.all/00/54/24140054.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR