A-1155463 ≥98% (by HPLC)
|
Warning
|
Synonyms:
2-(8-(benzo[d]thiazol-2-ylcarbamoyl)-3,4-dihydroisoquinolin-2(1H)-yl)-5-(3-(4-(3-(dimethylamino)prop-1-yn-1-yl)-2-fluorophenoxy)propyl)thiazole-4-carboxylic acid
B1821-5
B1821-25
76270-780EA
175.12
USD
76270-780
76270-782
A-1155463 ≥98% (by HPLC)
A-1155463
A-1155463 is a highly potent and selective BCL-XL inhibitor. A-1155463 is more potent against BCL-XL-dependent cell lines relative to WEHI-539 (Cat. No. B1819), while possessing none of its inherent pharmaceutical liabilities.
Formula:
C₃₅H₃₂FN₅O₄S₂ MW: 669,79 g/mol |
MDL Number:
MFCD29924714 CAS Number: 1235034-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...