CGM097 ≥98% (by HPLC)

Synonyms: (S)-1-(4-chlorophenyl)-7-isopropoxy-6-methoxy-2-(4-(methyl(((1r,4S)-4-(4-methyl-3-oxopiperazin-1-yl)cyclohexyl)methyl)amino)phenyl)-1,2-dihydroisoquinolin-3(4H)-one(S)-1-(4-Chlorophenyl)-7-isopropoxy-6-methoxy-2-(4-(methyl(((1r,4S)-4-(4-methyl-3-oxopiperazin-1-yl)cyclohexyl)methyl)amino)phenyl)-1,4-dihydroisoquinolin-3(2H)-one

B1820-1 B1820-5
76270-778EA 630.45 USD
76270-778 76270-776
CGM097 ≥98% (by HPLC)
CGM097

CGM097 is a potent and selective MDM2 inhibitor; an orally bioavailable HDM2 (human homolog of double minute 2) antagonist with potential antineoplastic activity.

Formula: C₃₈H₄₇ClN₄O₄
MW: 659,26 g/mol
MDL Number: MFCD28144684
CAS Number: 1313363-54-0
https://us.vwr.com/stibo/web/std.lang.all/00/52/24140052.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR