Luminol (3-Aminophthalhydrazide)

Synonyms: 5-Amino-2,3-dihydrophthalazine-1,4-dioneNSC 50645-Amino-2,3-Dihydro-1,4-Phthalazinedione

B1807-5G B1807-1G
76270-754EA 262.69 USD
76270-754 76270-752
Luminol (3-Aminophthalhydrazide)
Luminol (3-Aminophthalhydrazide)

Luminol is a useful probe that displays chemiluminescence upon oxidation.  Horseradish peroxidase (HRP), combined with hydrogen peroxide as an activator, causes luminescent peroxidation of luminol. Luminol can also be oxidized, and chemiluminesce, by compounds containing iron, copper, gold, and cyanide.

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD00006890
CAS Number: 521-31-3
https://us.vwr.com/stibo/web/std.lang.all/00/32/24140032.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR