4-Methylumbelliferyl-alpha-D-glucopyranoside ≥98%

Synonyms: 4-MU-alpha-D-Glc

B1805-250 B1805-50
76270-746EA 437.81 USD
76270-746 76270-744
4-Methylumbelliferyl-alpha-D-glucopyranoside ≥98%
4-Methylumbelliferyl-alpha-D-glucopyranoside

4-Methylumbelliferyl-alpha-D-glucopyranoside is a useful fluorogenic substrate for alpha-D-glucosidase.

Formula: C₁₆H₁₈O₈
MW: 338,32 g/mol
MDL Number: MFCD00067661
CAS Number: 17833-43-1
https://us.vwr.com/stibo/web/std.lang.all/00/29/24140029.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR