5-Bromo-4-chloro-3-indoxyl-N-acetyl-beta-D-glucosaminide ≥98%

Warning

Synonyms: X-beta-D-GlcNAc5-Bromo-4-chloro-3-indoxyl-2-acetamido-2-deoxy-beta-D-glucopyranoside

B1803-1000 B1803-250
76270-738EA 321.07 USD
76270-738 76270-736
5-Bromo-4-chloro-3-indoxyl-N-acetyl-beta-D-glucosaminide ≥98%
5-Bromo-4-chloro-3-indoxyl-N-acetyl-beta-D-glucosaminide

5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-glucosaminide is a histochemical substrate for N-Acetylglucosaminidase. It releases an insoluble blue chromophore after enzymatic action.

Formula: C₁₆H₁₈BrClN₂O₆
MW: 449,69 g/mol
MDL Number: MFCD00040631
CAS Number: 4264-82-8
https://us.vwr.com/stibo/web/std.lang.all/00/26/24140026.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR