D-Luciferin-6-O-β-D-galactopyranoside
B1802-25
B1802-5
76270-734EA
904.81
USD
76270-734
76270-732
D-Luciferin-6-O-β-D-galactopyranoside
D-Luciferin-6-O-β-D-galactopyranoside
Luciferin-β-galactoside is a substrate for the commonly used reporter enzyme β-galactosidase. The substrate is cleaved by β-galactosidase to form luciferin and galactose. When used in a model system expressing firefly luciferase, the luciferin is then utilized in a firefly luciferase reaction to generate light.
Formula:
C₂₂H₂₄O₇ |
MDL Number:
MFCD09038345 CAS Number: 131474-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...