Lucigenin

Synonyms: 10,10'-Dimethyl-9,9'-biacridinium dinitrateN,N′-Dimethyl-9,9′-biacridinium dinitrateBis(N-methylacridinium) nitrate

B1801-100 B1801-500
76270-728EA 88.63 USD
76270-728 76270-730
Lucigenin
Lucigenin

Lucigenin is a chemiluminescent probe for the detection of superoxide in biological systems. It has been used for the generation of chemiluminescence in the presence of a reductant and oxygen. Lucigenin is also used as a fluorescent chloride-sensitive indicator, with fluorescence being quenched by chloride (excitation: 455 nm; emission: 505 nm). Lucigenin fluorescence is insensitive to phosphate, sulfate, and nitrate.

Formula: C₂₈H₂₂N₄O₆
MW: 510.51 g/mol
MDL Number: MFCD00011925
CAS Number: 2315-97-1
https://us.vwr.com/stibo/web/std.lang.all/00/24/24140024.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR