CCT128930 ≥98%

Synonyms: 4-(4-chlorobenzyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-amineCCT128930CCT-1289304-[(4-chlorophenyl)methyl]-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-amineCCT 128930

B1763-1 B1763-5
76269-210EA 145.93 USD
76269-210 76269-212
CCT128930 ≥98%
CCT128930

CCT128930 is a potent, selective, novel and ATP-competitive inhibitor for AKT2 (IC50= 6 nM). It also has 28-fold and 20-fold selectivity over PKA kinase (IC50= 168 nM) and p70S6K (IC50= 120 nM), respectively.

Formula: C₁₈H₂₀ClN₅
MW: 341,84 g/mol
MDL Number: MFCD13177916
CAS Number: 885499-61-6
https://us.vwr.com/stibo/web/std.lang.all/00/18/24140018.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR