WAY-600 ≥98%

Synonyms: WAY6004-[6-(1H-indol-5-yl)-1-[1-(pyridin-3-ylmethyl)piperidin-4-yl]pyrazolo[3,4-d]pyrimidin-4-yl]morpholineWAY-600

B1731-1 B1731-5
76268-564EA 112.26 USD
76268-564 76268-854
WAY-600 ≥98%
WAY-600

WAY-600 is a potent, ATP-competitive and selective inhibitor of mTOR with IC50 of 9 nM; blocks mTORC1/P-S6K(T389) and mTORC2/P-AKT(S473) but not P-AKT(T308); selective for mTOR than PI3Kα (>100-fold) and PI3Kγ (>500-fold).

Formula: C₂₈H₃₀N₈O
MW: 494,59 g/mol
MDL Number: MFCD22419019
CAS Number: 1062159-35-6
https://us.vwr.com/stibo/web/std.lang.all/00/07/24140007.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR