VE-821 ≥98%

Synonyms: VE8213-amino-6-(4-methylsulphonylphenyl)-N-phenylpyrazine-2-carboxamideVE 821VE-8213-Amino-6-(4-(methylsulfonyl)phenyl)-N-phenylpyrazine-2-carboxamide3-amino-6-(4-methylsulfonylphenyl)-N-phenylpyrazine-2-carboxamide3-Amino-6-(4-(methylsulphonyl)phenyl)-N-phenylpyrazine-2-carboxamide

B1723-5 B1723-25
76268-538EA 624.61 USD
76268-538 76268-536
VE-821 ≥98%
VE-821

VE-821 is a potent and selective ATP competitive inhibitor of ATR with Ki/IC50 of 13 nM/26 nM.

Formula: C₁₈H₁₆N₄O₃S
MW: 368,41 g/mol
MDL Number: MFCD19443686
CAS Number: 1232410-49-9
https://us.vwr.com/stibo/web/std.lang.all/99/96/24139996.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR