CP-466722 ≥98%

Warning

Synonyms: 2-(6,7-dimethoxyquinazolin-4-yl)-5-pyridin-2-yl-1,2,4-triazol-3-amineCP 466722CP-466722CP466722

B1722-5 B1722-1
76268-534EA 321.07 USD
76268-534 76268-532
CP-466722 ≥98%
CP-466722

CP-466722 is a selective inhibitor of ATM kinase with IC50 value of 0.41 μM.

Formula: C₁₇H₁₅N₇O₂
MW: 349,35 g/mol
MDL Number: MFCD17392566
CAS Number: 1080622-86-1
https://us.vwr.com/stibo/web/std.lang.all/99/95/24139995.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR